2-Mercaptobenzoxazole, 98+%, Thermo Scientific Chemicals
2-Mercaptobenzoxazole, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Mercaptobenzoxazole, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number A12346.22
also known as A12346-22
Price (USD)/ Each
134.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
134.00
Add to cart
2-Mercaptobenzoxazole, 98+%, Thermo Scientific Chemicals
Catalog numberA12346.22
Price (USD)/ Each
134.00
-
Add to cart
Chemical Identifiers
CAS2382-96-9
IUPAC Name2,3-dihydro-1,3-benzoxazole-2-thione
Molecular FormulaC7H5NOS
InChI KeyFLFWJIBUZQARMD-UHFFFAOYSA-N
SMILESS=C1NC2=CC=CC=C2O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to dark cream to pale brown or grey
FormPowder
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0% (non-U.S. specification)
Assay (HPLC)≥98.0% (non-U.S. specification)
Assay from Suppliers CofA≥98.0% (HPLC, U.S. specification)
View more
2-Mercaptobenzoxazole is used in the surface treatment of materials. It is utilized for protection of metals from corrosion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Mercaptobenzoxazole is used in the surface treatment of materials. It is utilized for protection of metals from corrosion.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Successive alkylation with different alkyl halides provides a route to unsymmmetrical dialkyl sulfides: J. Org. Chem., 48, 3842 (1983):
  2. If the two alkyl groups are identical the reaction can be run as a one-pot sequence.
  3. Ziemkowska, W.; Skałkowska, J.; Ochal, Z.; Cyrański, M. K.; Dobrzycki, Ł.; Madura, I.; Zachara, J. Role of Lewis bases in reactions of aluminum and gallium trialkyls with 2-mercaptobenzoxazole. J. Organomet. Chem. 2015, 776, 1-6.
  4. Gao, Y. Z.; Zhang, Y. A.; Zhang, J. Synthesis, structures, and magnetic properties of tetranuclear nickel and cobalt complexes with 2-mercaptobenzoxazole. Inorg. Chem. Commun. 2015, 54, 85-88.