Thermo Scientific Chemicals

1-Bromo-2-chloroethane, 98%, Thermo Scientific Chemicals

Catalog number: A12349.36
500 g, Each
Thermo Scientific Chemicals

1-Bromo-2-chloroethane, 98%, Thermo Scientific Chemicals

Catalog number: A12349.36
500 g, Each
Quantity
Catalog number: A12349.36
also known as A12349-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
107-04-0
IUPAC Name
1-bromo-2-chloroethane
Molecular Formula
C2H4BrCl
InChI Key
IBYHHJPAARCAIE-UHFFFAOYSA-N
SMILES
ClCCBr
Form
Liquid
Identification (FTIR)
Conforms
Refractive Index
1.4880-1.4920 @ 20?C
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%

Description

1-Bromo-2-chloroethane was used in regio- and diastereoselective synthesis of 2-alkylidenetetrahydrofurans. 1-Bromo-2-chloroethane is used as a solvent and as an intermediate for organic synthesis. It is used as a as fumigant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2-chloroethane was used in regio- and diastereoselective synthesis of 2-alkylidenetetrahydrofurans. 1-Bromo-2-chloroethane is used as a solvent and as an intermediate for organic synthesis. It is used as a as fumigant.

Solubility
Soluble in water ( 7 g/L (20°C). Soluble in ethanol, ethyl ether, chloroform.

Notes
Light Sensitive. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text