Used as a protein denaturantUrea is a raw fabric for the manufacture of organic resins and is used as protein denaturant sealant and in agriculture, dyes, fuels, functional fluids, intermediates, paint additives and coating additives. Urea is used as a nitrogen-release fertilizer, on-corroding alternative to rock salt for road de-icing, browning agent in factory-produced pretzels, cloud seeding agent, a deep eutectic solvent and type of ionic liquid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used as a protein denaturantUrea is a raw fabric for the manufacture of organic resins and is used as protein denaturant sealant and in agriculture, dyes, fuels, functional fluids, intermediates, paint additives and coating additives. Urea is used as a nitrogen-release fertilizer, on-corroding alternative to rock salt for road de-icing, browning agent in factory-produced pretzels, cloud seeding agent, a deep eutectic solvent and type of ionic liquid.
Solubility
Soluble in water, alcohol, methanol, glycerol, benzene and concentrated hydrochloric acid. Practically insoluble in chloroform and ether.
Notes
Stable at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
- Widely used in diazotization reactions for the destruction of excess HNO2.
- For the one-pot conversion of carboxylic acids to nitriles, by heating with urea in the presence of 1.5-2.0 moles of sulfamic acid, see: Chimia, 25, 94 (1971).
- For a procedure for conversion of carboxylic acids to primary amides by heating with urea, see: Org. Synth. Coll., 3, 768 (1955); 4, 513 (1963).
- Reacts with alcohols on prolonged heating to give alkyl carbamates; see, e.g.: Org. Synth. Coll., 1, 140 (1941).
- Zhang, Y.; Han, C.; Nadagouda, M. N.; Dionysiou, D. D. The fabrication of innovative single crystal N, F-codoped titanium dioxide nanowires with enhanced photocatalytic activity for degradation of atrazine. Appl. Catal. B 2015, 168-169, 550-558.
- Amendola, V.; Fabbrizzi, L.; Mosca, L. Anion recognition by hydrogen bonding: urea-based receptors. Chem. Soc. Rev. 2010, 39, 3889-3915.