Thermo Scientific Chemicals

Diphenyl carbonate, 99%, Thermo Scientific Chemicals

Catalog number: A12362.0I
5000 g, Each
Thermo Scientific Chemicals

Diphenyl carbonate, 99%, Thermo Scientific Chemicals

Catalog number: A12362.0I
5000 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A12362.0I
also known as A12362-0I
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
102-09-0
IUPAC Name
diphenyl carbonate
Molecular Formula
C13H10O3
InChI Key
ROORDVPLFPIABK-UHFFFAOYSA-N
SMILES
O=C(OC1=CC=CC=C1)OC1=CC=CC=C1
Identification (FTIR)
Conforms
Melting Point (clear melt)
75.0-83.0?C
Form
Flakes or granules
Assay (GC)
≥98.5%
Appearance (Color)
White

Description

Used for synthesis of many important organic compounds. As agrochemical intermediates, Dyestuff intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used for synthesis of many important organic compounds. As agrochemical intermediates, Dyestuff intermediate.

Solubility
Soluble in ethanol, diethyl ether, carbon tetrachloride, acetic acid. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text