4-Methoxy-o-phenylenediamine dihydrochloride, 96%, Thermo Scientific Chemicals
4-Methoxy-o-phenylenediamine dihydrochloride, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methoxy-o-phenylenediamine dihydrochloride, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number A12363.14
also known as A12363-14
Price (USD)/ Each
356.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
356.00
Add to cart
4-Methoxy-o-phenylenediamine dihydrochloride, 96%, Thermo Scientific Chemicals
Catalog numberA12363.14
Price (USD)/ Each
356.00
-
Add to cart
Chemical Identifiers
CAS59548-39-9
IUPAC Name4-methoxybenzene-1,2-diamine hydrochloridyl
Molecular FormulaC7H10ClN2O
InChI KeyNOFDEJFDTTYTCG-UHFFFAOYSA-N
SMILES[Cl].COC1=CC=C(N)C(N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Purple to pink
Assay (Titration ex Chloride)≥95.0 to ≤105.0%
4-Methoxy-o-phenylenediamine is widely used as an intermediate in organic synthesis and pharmaceuticals. It is an important precursor for the synthesis of other heterocyclic compounds. It is used in the determination of glyoxal, methylglyoxal, and diacetyl in urine using 4-methoxy-o-phenylenediamine as derivatizing reagent by High-performance liquid chromatography (HPLC).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxy-o-phenylenediamine is widely used as an intermediate in organic synthesis and pharmaceuticals. It is an important precursor for the synthesis of other heterocyclic compounds. It is used in the determination of glyoxal, methylglyoxal, and diacetyl in urine using 4-methoxy-o-phenylenediamine as derivatizing reagent by High-performance liquid chromatography (HPLC).

Solubility
Soluble in water.

Notes
Store in a cool place. Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents. Hygroscopic.
RUO – Research Use Only

General References:

  1. Escobosa, A. R. C.; Ojeda, A. G.; Wrobel, K.; Magana, A. A.; Wrobel, K. Methylglyoxal is associated with bacteriostatic activity of high fructose agave syrups. Food Chem. 2014, 165, 444-450.
  2. Ojeda, A. G.; Escobosa, A. R. C.; Wrobel, K.; Barrientos, E. Y.; Wrobel, K. Effect of Cd(II) and Se(IV) exposure on cellular distribution of both elements and concentration levels of glyoxal and methylglyoxal in Lepidium sativum. Metallomics 2013, 5 (9), 1254-1261.