Thermo Scientific Chemicals

2,4-Dichlorobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A12372.0B
1000 g, Each
Thermo Scientific Chemicals

2,4-Dichlorobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A12372.0B
1000 g, Each
Quantity
Catalog number: A12372.0B
also known as A12372-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
50-84-0
IUPAC Name
2,4-dichlorobenzoic acid
Molecular Formula
C7H4Cl2O2
InChI Key
ATCRIUVQKHMXSH-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C(Cl)C=C1Cl
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Form
Powder or lumps
Assay (Silylated GC)
≥97.5%
Melting Point (clear melt)
160.0-166.0?C
Appearance (Color)
White

Description

2,4-Dichlorobenzoic acid is used as reagent during the synthesis of pyrimido[2?,1?:2,3]thiazolo[4,5-b]quinoxaline derivatives. It is also used as starting reagent during the synthesis of 1-(substituted)-1,4-dihydro-6-nitro-4-oxo-7-(sub-secondary amino)-quinoline-3-carboxylic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dichlorobenzoic acid is used as reagent during the synthesis of pyrimido[2′,1′:2,3]thiazolo[4,5-b]quinoxaline derivatives. It is also used as starting reagent during the synthesis of 1-(substituted)-1,4-dihydro-6-nitro-4-oxo-7-(sub-secondary amino)-quinoline-3-carboxylic acids.

Solubility
Soluble in ethanol (100 mg/ml), water (0.48 mg/ml), alcohol, ether, and acetone.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text