Thermo Scientific Chemicals

2-Aminopyridine, 99%, Thermo Scientific Chemicals

Catalog number: A12374.14
25 g, Each
Thermo Scientific Chemicals

2-Aminopyridine, 99%, Thermo Scientific Chemicals

Catalog number: A12374.14
25 g, Each
Quantity
Catalog number: A12374.14
also known as A12374-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
504-29-0
IUPAC Name
pyridin-2-amine
Molecular Formula
C5H6N2
InChI Key
ICSNLGPSRYBMBD-UHFFFAOYSA-N
SMILES
NC1=CC=CC=N1
Identification (FTIR)
Conforms
Form
Crystalline powder, crystals, flakes or lumps
Melting Point (clear melt)
55-60?C
Appearance (Color)
White to cream to yellow
Assay (GC)
≥98.5%

Description

A compound has been shown to reversibly block voltage-dependent potassium channels. Also a common impurity from the synthesis of compounds found in hair dye. Used as a water miscible solvent and as an intermediate in manufacturing pharmaceuticals like diuretics. It is used as a research tool, in characterizing subtypes of the potassium channel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A compound has been shown to reversibly block voltage-dependent potassium channels. Also a common impurity from the synthesis of compounds found in hair dye. Used as a water miscible solvent and as an intermediate in manufacturing pharmaceuticals like diuretics. It is used as a research tool, in characterizing subtypes of the potassium channel.

Solubility
Slightly soluble in water(8g/100 mL at 19°C)

Notes
Hygroscopic. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text