Thermo Scientific Chemicals

tert-Butylbenzene, 99%, Thermo Scientific Chemicals

Catalog number: A12378.AK
250 mL, Each
Thermo Scientific Chemicals

tert-Butylbenzene, 99%, Thermo Scientific Chemicals

Catalog number: A12378.AK
250 mL, Each
Quantity
Catalog number: A12378.AK
also known as A12378-AK
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
98-06-6
IUPAC Name
tert-butylbenzene
Molecular Formula
C10H14
InChI Key
YTZKOQUCBOVLHL-UHFFFAOYSA-N
SMILES
CC(C)(C)C1=CC=CC=C1
Appearance (Color)
Clear colorless
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Refractive Index
1.4910-1.4940 @ 20?C
Form
Liquid

Description

tert-Butylbenzene is an useful intermediate for manufacturing agrochemicals and pharmaceuticals. tert-Butyl Toluene is used as a solvent for resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butylbenzene is an useful intermediate for manufacturing agrochemicals and pharmaceuticals. tert-Butyl Toluene is used as a solvent for resins.

Solubility
Insoluble in water(0.03g/100 mL at 20°C) .

Notes
Keep container tightly closed in a cool, dry and well-ventilated place. Store away from strong oxidizing agents, acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text