Thermo Scientific Chemicals

Sodium chloroacetate, 98%, Thermo Scientific Chemicals

Catalog number: A12379.36
500 g, Each
Thermo Scientific Chemicals

Sodium chloroacetate, 98%, Thermo Scientific Chemicals

Catalog number: A12379.36
500 g, Each
Quantity
Catalog number: A12379.36
also known as A12379-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3926-62-3
IUPAC Name
sodium 2-chloroacetate
Molecular Formula
C2H2ClNaO2
InChI Key
FDRCDNZGSXJAFP-UHFFFAOYSA-M
SMILES
[Na+].[O-]C(=O)CCl
Appearance (Color)
White
Water Content (Karl Fischer Titration)
≤1.0%
Form
Crystals or powder or crystalline powder or compacted powder or granules or flakes or lumps
Identification (FTIR)
Conforms
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%

Description

Sodium chloroacetate is used to prepare dyes and active pharmaceutical ingredients. It is also used as an odor agent, surface active agent and viscosity adjustor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium chloroacetate is used to prepare dyes and active pharmaceutical ingredients. It is also used as an odor agent, surface active agent and viscosity adjustor.

Solubility
Soluble in water, ether, chloroform, benzene and alcohol.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text