Thermo Scientific Chemicals

(S)-(-)-Nicotine, 99%, Thermo Scientific Chemicals

Catalog number: A12398.22
100 g, Each
Thermo Scientific Chemicals

(S)-(-)-Nicotine, 99%, Thermo Scientific Chemicals

Catalog number: A12398.22
100 g, Each
Quantity
Catalog number: A12398.22
also known as A12398-22
Price (USD)
Have Questions?

Chemical Identifiers

CAS
54-11-5
IUPAC Name
3-[(2S)-1-methylpyrrolidin-2-yl]pyridine
Molecular Formula
C10H14N2
InChI Key
SNICXCGAKADSCV-JTQLQIEISA-N
SMILES
CN1CCC[C@H]1C1=CC=CN=C1
Optical Rotation
-165 ? 5? (neat) or -146 ? 6? (c=2, ethanol)
Appearance (Color)
Clear colorless to yellow to orange to brown
Form
Liquid
Assay (GC)
≥98.5%
Refractive Index
1.5250-1.5290 @ 20?C

Description

(S)-(-)-Nicotine is utilized as a therapeutic agent to eliminate smoking with the damage it does to health. It is widely used as a drug to enhance the memory or other cognition functions, which is second to caffeine. Lesser levels of nicotine given in the form of gum, may help to eliminate smoking.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description
• L-Nicotine is a naturally occuring alkaloid in the nightshade family of plants
• It is also known as 3-(1-methyl-2-pyrrolidinyl)pyridine or (S)-(-)-nicotine
Applications
• L-Nicotine has been shown in studies to alleviate smoking habits
• It has also been shown to enhance cognitive functions such as memory
• Studies show that lower levels of nicotine in the form of gum may also help eliminate smoking habits

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text