S-Methyl thioacetate, 98+%, Thermo Scientific Chemicals
S-Methyl thioacetate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

S-Methyl thioacetate, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number A12400.14
also known as A12400-14
Price (USD)/ Each
256.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
256.00
Add to cart
S-Methyl thioacetate, 98+%, Thermo Scientific Chemicals
Catalog numberA12400.14
Price (USD)/ Each
256.00
-
Add to cart
Chemical Identifiers
CAS1534-08-3
IUPAC Name1-(methylsulfanyl)ethan-1-one
Molecular FormulaC3H6OS
InChI KeyOATSQCXMYKYFQO-UHFFFAOYSA-N
SMILESCSC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥98.0%
FormLiquid
Refractive Index1.4615-1.4655 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only