Thermo Scientific Chemicals

Ethyl 3-bromopropionate, 99%, Thermo Scientific Chemicals

Catalog number: A12402.22
100 g, Each
Thermo Scientific Chemicals

Ethyl 3-bromopropionate, 99%, Thermo Scientific Chemicals

Catalog number: A12402.22
100 g, Each
Quantity
Catalog number: A12402.22
also known as A12402-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
539-74-2
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.5%
Form
Liquid
Identification (FTIR)
Conforms
Refractive Index
1.4510-1.4540 @ 20?C

Description

Ethyl 3-bromopropionate was used in the preparation of (3?,4?-dimethoxyphenyl)-5-oxopentanoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-bromopropionate was used in the preparation of (3′,4′-dimethoxyphenyl)-5-oxopentanoic acid.

Solubility
Soluble in water, alcohol, chloroform, benzene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, Bases, Reducing agents
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text