Thermo Scientific Chemicals

Taurine, 99%, Thermo Scientific Chemicals

Catalog number: A12403.22
100 g, Each
Thermo Scientific Chemicals

Taurine, 99%, Thermo Scientific Chemicals

Catalog number: A12403.22
100 g, Each
Quantity
Catalog number: A12403.22
also known as A12403-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
107-35-7
IUPAC Name
2-aminoethane-1-sulfonic acid
Molecular Formula
C2H7NO3S
InChI Key
XOAAWQZATWQOTB-UHFFFAOYSA-N
SMILES
NCCS(O)(=O)=O
Appearance (Color)
White
Form
Crystals or crystalline powder or needles
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5% (non-U.S. specification)
Assay from Suppliers CofA
≥98.5% (U.S. specification)
Comment
Specification differs for U.S. and non-U.S. material where indicated

Description

Taurine plays an important role in cardiovascular function, skeletal muscle function, the retina and the central nervous system. It acts as an antioxidant as well as protecting against the toxicity of various substances such as lead and cadmium. It is also used in contact lens solution and an ingredient in numerous energy drinks.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Taurine plays an important role in cardiovascular function, skeletal muscle function, the retina and the central nervous system. It acts as an antioxidant as well as protecting against the toxicity of various substances such as lead and cadmium. It is also used in contact lens solution and an ingredient in numerous energy drinks.

Solubility
Soluble in water and dimethyl sulfoxide. Insoluble in absolute ethanol and diethyl ether.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text