Thermo Scientific Chemicals

Methyl benzoate, 99%, Thermo Scientific Chemicals

Catalog number: A12404.30
250 g, Each
Thermo Scientific Chemicals

Methyl benzoate, 99%, Thermo Scientific Chemicals

Catalog number: A12404.30
250 g, Each
Quantity
Catalog number: A12404.30
also known as A12404-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
93-58-3
IUPAC Name
methyl benzoate
Molecular Formula
C8H8O2
InChI Key
QPJVMBTYPHYUOC-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=CC=C1
Appearance (Color)
Clear colorless
Identification (FTIR)
Conforms
Form
Liquid
Assay (GC)
≥98.5%
Refractive Index
1.5155-1.5185 @ 20?C

Description

Methyl benzoate is used in perfumery. It also finds use as a solvent. It acts as an intermediate and odor agents. Further, it is used to attract insects such as orchid bees. It is also used for cellulose esters, cellulose ethers, synthetic resin and rubber solvent and polyester fibers to assist in the preparation of flavor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl benzoate is used in perfumery. It also finds use as a solvent. It acts as an intermediate and odor agents. Further, it is used to attract insects such as orchid bees. It is also used for cellulose esters, cellulose ethers, synthetic resin and rubber solvent and polyester fibers to assist in the preparation of flavor.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text