Thermo Scientific Chemicals

Iodoacetic acid, 97+%, Thermo Scientific Chemicals

Catalog number: A12410.14
25 g, Each
Thermo Scientific Chemicals

Iodoacetic acid, 97+%, Thermo Scientific Chemicals

Catalog number: A12410.14
25 g, Each
Quantity
Catalog number: A12410.14
also known as A12410-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
64-69-7
IUPAC Name
2-iodoacetic acid
Molecular Formula
C2H3IO2
InChI Key
JDNTWHVOXJZDSN-UHFFFAOYSA-N
SMILES
OC(=O)CI
Assay (Aqueous acid-base Titration)
≥97.0 to ≤103.0%
Melting Point (clear melt)
74-85?C
Appearance (Color)
White to yellow to orange-brown or pale cream to cream
Form
Powder or crystalline powder or crystals or flakes
Identification (FTIR)
Conforms

Description

Iodoacetic acid is used as a reagent for the modification of sulfhydryl groups in organic synthesis. It reacts with cysteine moiety in proteins to prevent the re-formation of disulfide bonds during protein sequencing. It is also used as a general reagent in acylation reactions as well as in glycopeptide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodoacetic acid is used as a reagent for the modification of sulfhydryl groups in organic synthesis. It reacts with cysteine moiety in proteins to prevent the re-formation of disulfide bonds during protein sequencing. It is also used as a general reagent in acylation reactions as well as in glycopeptide synthesis.

Solubility
Soluble in water, alcohol and ethanol. Slightly soluble in ether and chloroform.

Notes
Light sensitive. Store in cool place. Incompatible with strong oxidizing agents, bases and strong reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text