Thermo Scientific Chemicals

2-Bromobenzylamine, 96%, Thermo Scientific Chemicals

Catalog number: A12412.06
5 g, Each
Thermo Scientific Chemicals

2-Bromobenzylamine, 96%, Thermo Scientific Chemicals

Catalog number: A12412.06
5 g, Each
Quantity
Catalog number: A12412.06
also known as A12412-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3959-05-5
IUPAC Name
1-(2-bromophenyl)methanamine
Molecular Formula
C7H8BrN
InChI Key
NOYASZMZIBFFNZ-UHFFFAOYSA-N
SMILES
NCC1=CC=CC=C1Br
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Refractive Index
1.5880-1.6000 @ 20°C
Assay (GC)
≥95.0%

Description

Pharmaceutical intermediates. As intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pharmaceutical intermediates. As intermediate in organic synthesis.

Solubility
Insoluble in Water. Soluble in Alcohol and Ether.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, carbondioxide..
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text