2-Bromobenzylamine, 96%, Thermo Scientific Chemicals
2-Bromobenzylamine, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromobenzylamine, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number A12412.14
also known as A12412-14
Price (USD)/ Each
277.65
Online exclusive
308.00 
Save 30.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
277.65
Online exclusive
308.00 
Save 30.35 (10%)
Add to cart
2-Bromobenzylamine, 96%, Thermo Scientific Chemicals
Catalog numberA12412.14
Price (USD)/ Each
277.65
Online exclusive
308.00 
Save 30.35 (10%)
-
Add to cart
Chemical Identifiers
CAS3959-05-5
IUPAC Name1-(2-bromophenyl)methanamine
Molecular FormulaC7H8BrN
InChI KeyNOYASZMZIBFFNZ-UHFFFAOYSA-N
SMILESNCC1=CC=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Refractive Index1.5880-1.6000 @ 20°C
Assay (GC)≥95.0%
Pharmaceutical intermediates. As intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pharmaceutical intermediates. As intermediate in organic synthesis.

Solubility
Insoluble in Water. Soluble in Alcohol and Ether.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, carbondioxide..
RUO – Research Use Only

General References:

  1. Hilton ST, et al. A tandem radical approach to the ABCE-rings of the Aspidosperma and Strychnos alkaloids. Chem. Commun. (Camb.) 2001, 2209-210.
  2. S B Ross. Long-term effects of N-2-chlorethyl-N-ethyl-2-bromobenzylamine hydrochloride on noradrenergic neurones in the rat brain and heart.. Br J Pharmacol. 1976, 58(4),521-527.