Thermo Scientific Chemicals

Sodium cyanate, 95%, Thermo Scientific Chemicals

Catalog number: A12413.22
100 g, Each
Thermo Scientific Chemicals

Sodium cyanate, 95%, Thermo Scientific Chemicals

Catalog number: A12413.22
100 g, Each
Quantity
Catalog number: A12413.22
also known as A12413-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
917-61-3
IUPAC Name
sodium cyanate
Molecular Formula
CNNaO
InChI Key
ZVCDLGYNFYZZOK-UHFFFAOYSA-M
SMILES
[Na+].[O-]C#N
Appearance (Color)
White to pale cream
Form
Powder
Assay (ex Sulphated Ash)
≥94.0%
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
≤3.5%

Description

Sodium cyanate is an ideal nucleophile, and these nucleophilic properties make it a major contributor to the stereospecificity in certain reactions such as in the production of chiral oxazolidone. Sodium cyanate is a useful reagent in producing asymmetrical urea that have a range of biological activity mostly in aryl isocyanate intermediates. Such intermediates as well as sodium cyanate have been used in medicine as a means of counterbalancing carcinogenic effects on the body.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium cyanate is an ideal nucleophile, and these nucleophilic properties make it a major contributor to the stereospecificity in certain reactions such as in the production of chiral oxazolidone. Sodium cyanate is a useful reagent in producing asymmetrical urea that have a range of biological activity mostly in aryl isocyanate intermediates. Such intermediates as well as sodium cyanate have been used in medicine as a means of counterbalancing carcinogenic effects on the body.

Solubility
Soluble in water 11.6 g/100 mL (25°C). Slightly soluble in ammonia, benzene. Insoluble in diethyl ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text