Thermo Scientific Chemicals

4-Isopropylphenylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A12416.06
5 g, Each
Thermo Scientific Chemicals

4-Isopropylphenylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A12416.06
5 g, Each
Quantity
Catalog number: A12416.06
also known as A12416-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
118427-29-5
IUPAC Name
[4-(propan-2-yl)phenyl]hydrazine
Molecular Formula
C9H14N2
InChI Key
ZYATZFJUOXJFPY-UHFFFAOYSA-N
SMILES
CC(C)C1=CC=C(NN)C=C1
Appearance (Color)
White to cream to brown or yellow
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%
Form
Powder or flakes

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text