A reagent used for deriving primary amines. 2,3,4,5,6-Pentafluorobenzoyl chloride was used for derivatization of 5-androstenediol, testosterone, estradiol , dihydrotestosterone and estrone in a study to determine their serum levels. 2,3,4,5,6-Pentafluorobenzoyl chloride was used to derivative dimethylamine(DMA) in study to develop a rapid and accurate GC-MS method for determination of DMA in human urine. It was used in a study to detect the presence of Enterobacter cloacae in cultures of Leuconostoc mesenteroide by gas chromatography/mass spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
A reagent used for deriving primary amines. 2,3,4,5,6-Pentafluorobenzoyl chloride was used for derivatization of 5-androstenediol, testosterone, estradiol , dihydrotestosterone and estrone in a study to determine their serum levels. 2,3,4,5,6-Pentafluorobenzoyl chloride was used to derivative dimethylamine(DMA) in study to develop a rapid and accurate GC-MS method for determination of DMA in human urine. It was used in a study to detect the presence of Enterobacter cloacae in cultures of Leuconostoc mesenteroide by gas chromatography/mass spectrometry.
Solubility
Decomposes in water.
Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, Water, Alcohols, Strong bases.
RUO – Research Use Only
General References:
- Frank Giton et. al. Serum sex steroids measured in middle-aged European and African-Caribbean men by gas chromatography-mass spectrometry. European Journal of Endocrinology. 2011, 165 (6),917-924.
- Dimitrios Tsikas et. al. Accurate quantification of dimethylamine (DMA) in human urine by gas chromatography-mass spectrometry as pentafluorobenzamide derivative: evaluation of the relationship between DMA and its precursor asymmetric dimethylarginine (ADMA) in health and disease.. Journal of Chromatography B.2007, 851 (1-2),229-239.
- Reagent for the preparation of derivatives of primary amines, etc. for GC. For monographs on GC reagents, see: D. R. Knapp, Handbook of Analytical Derivatisation Reactions, Wiley, N.Y. (1979); Handbook of Derivatives for Chromatography, 2nd ed., K. Blau and J. M. Halket, Eds., Wiley, Chichester (1993).