Thermo Scientific Chemicals

4-(Methylamino)benzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: A12426.06
5 g, Each
Thermo Scientific Chemicals

4-(Methylamino)benzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: A12426.06
5 g, Each
Quantity
Catalog number: A12426.06
also known as A12426-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10541-83-0
IUPAC Name
4-(methylamino)benzoic acid
Molecular Formula
C8H9NO2
InChI Key
ZVIDMSBTYRSMAR-UHFFFAOYSA-N
SMILES
CNC1=CC=C(C=C1)C(O)=O
Form
Powder
Assay (HPLC)
≥96.0%
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Melting Point (clear melt)
156.0-166.0?C
Appearance (Color)
Pale cream to cream

Description

4-(Methylamino)benzoic acid important for the preparation of other pharmaceutical products, dyes, flavors, and preservatives. .For the preparation of medicines and other fine chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Methylamino)benzoic acid important for the preparation of other pharmaceutical products, dyes, flavors, and preservatives. .For the preparation of medicines and other fine chemicals.

Solubility
Slightly soluble. Soluble in alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text