Thermo Scientific Chemicals

Potassium hydrogen carbonate, 99%, Thermo Scientific Chemicals

Catalog number: A12429.0E
2500 g, Each
Thermo Scientific Chemicals

Potassium hydrogen carbonate, 99%, Thermo Scientific Chemicals

Catalog number: A12429.0E
2500 g, Each
Quantity
Catalog number: A12429.0E
also known as A12429-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
298-14-6
IUPAC Name
potassium hydrogen carbonate
Molecular Formula
CHKO3
InChI Key
TYJJADVDDVDEDZ-UHFFFAOYSA-M
SMILES
[K+].OC([O-])=O
Identification (FTIR)
Conforms
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Form
Crystals or powder or crystalline powder
Appearance (Color)
White

Description

Potassium hydrogen carbonate is used in foods to regulate pH, as a fire suppression agent (BC dry chemical) and as a buffering agent in medications. It acts as an additive in winemaking and is an active component in club soda. Furthermore, it finds use in crops for the neutralization of acidic soils. It is also used as a source of carbon dioxide for leavening in baking. It is utilized to inhibit the growth of aspergillus parasiticus in czapek's agar. It is employed as a catalyst in synthetic fiber polymerization and olefin dehydrogenation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium hydrogen carbonate is used in foods to regulate pH, as a fire suppression agent (BC dry chemical) and as a buffering agent in medications. It acts as an additive in winemaking and is an active component in club soda. Furthermore, it finds use in crops for the neutralization of acidic soils. It is also used as a source of carbon dioxide for leavening in baking. It is utilized to inhibit the growth of aspergillus parasiticus in czapek′s agar. It is employed as a catalyst in synthetic fiber polymerization and olefin dehydrogenation.

Solubility
Soluble in water. Insoluble in alcohol.

Notes
Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text