Thermo Scientific Chemicals

Cyclobutanol, 96%, Thermo Scientific Chemicals

Catalog number: A12430.06
5 g, Each
Thermo Scientific Chemicals

Cyclobutanol, 96%, Thermo Scientific Chemicals

Catalog number: A12430.06
5 g, Each
Quantity
Catalog number: A12430.06
also known as A12430-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2919-23-5
IUPAC Name
cyclobutanol
Molecular Formula
C4H8O
InChI Key
KTHXBEHDVMTNOH-UHFFFAOYSA-N
SMILES
OC1CCC1
Form
Liquid
Refractive Index
1.4335-1.4395 @ 20?C
Appearance (Color)
Clear colorless
Assay (GC)
≥95.0%

Description

Has synthetic applications of gold-catalyzed ring expansions. Development of novel type of transition metal-catalyzed cascade reactions utilizing small ring systems and the applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Has synthetic applications of gold-catalyzed ring expansions. Development of novel type of transition metal-catalyzed cascade reactions utilizing small ring systems and the applications.

Solubility
Partly soluble.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acid chlorides, acid anhydrides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text