Has synthetic applications of gold-catalyzed ring expansions. Development of novel type of transition metal-catalyzed cascade reactions utilizing small ring systems and the applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Has synthetic applications of gold-catalyzed ring expansions. Development of novel type of transition metal-catalyzed cascade reactions utilizing small ring systems and the applications.
Solubility
Partly soluble.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acid chlorides, acid anhydrides.
RUO – Research Use Only
Jan. Rocek,; Annette E. Radkowsky.Mechanism of the chromic acid oxidation of cyclobutanol. J. Am. Chem. Soc. 1973 , 95(21),7123-7132.
Jan Rocek,; Annette E. Radkowsky.The role of chromium(IV) in oxidations by chromic acid. The oxidative cleavage of cyclobutanol. J. Am. Chem. Soc.1968 , 90(11), 2986-2988.