Thermo Scientific Chemicals

1,5-Dichloropentane, 98%, Thermo Scientific Chemicals

Catalog number: A12433.22
100 g, Each
Thermo Scientific Chemicals

1,5-Dichloropentane, 98%, Thermo Scientific Chemicals

Catalog number: A12433.22
100 g, Each
Quantity
Catalog number: A12433.22
also known as A12433-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
628-76-2
IUPAC Name
1,5-dichloropentane
Molecular Formula
C5H10Cl2
InChI Key
LBKDGROORAKTLC-UHFFFAOYSA-N
SMILES
ClCCCCCCl
Identification (FTIR)
Conforms
Refractive Index
1.4540-1.4580 @ 20?C
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥97.5%

Description

1,5-Dichloropentane is used in the preparation of symmetrical sulfide, tetrydrothiopyran (thiane) by reaction with sodium sulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Dichloropentane is used in the preparation of symmetrical sulfide, tetrydrothiopyran (thiane) by reaction with sodium sulfide.

Solubility
Miscible with ethanol, ether and chloroform. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text