Thermo Scientific Chemicals

Methyl 2-bromobenzoate, 99%, Thermo Scientific Chemicals

Catalog number: A12435.36
500 g, Each
Thermo Scientific Chemicals

Methyl 2-bromobenzoate, 99%, Thermo Scientific Chemicals

Catalog number: A12435.36
500 g, Each
Quantity
Catalog number: A12435.36
also known as A12435-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
610-94-6
IUPAC Name
methyl 2-bromobenzoate
Molecular Formula
C8H7BrO2
InChI Key
SWGQITQOBPXVRC-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=CC=C1Br
Appearance (Color)
Clear colorless
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Refractive Index
1.5575-1.5605 @ 20?C
Form
Liquid

Description

Methyl 2-bromobenzoate under Heck reaction with acrylonitrile results in lactonization to give the benzofuran-2-one 3-(cyanomethyl)-2-coumaranone. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-bromobenzoate under Heck reaction with acrylonitrile results in lactonization to give the benzofuran-2-one 3-(cyanomethyl)-2-coumaranone. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals

Solubility
Insoluble in water.

Notes
Store in a cool place. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text