Thermo Scientific Chemicals

1-Octyne, 98%, Thermo Scientific Chemicals

Catalog number: A12440.22
100 g, Each
Thermo Scientific Chemicals

1-Octyne, 98%, Thermo Scientific Chemicals

Catalog number: A12440.22
100 g, Each
Quantity
Catalog number: A12440.22
also known as A12440-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
629-05-0
IUPAC Name
oct-1-yne
Molecular Formula
C8H14
InChI Key
UMIPWJGWASORKV-UHFFFAOYSA-N
SMILES
CCCCCCC#C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Refractive Index
1.4145-1.4185 @ 20°C (non-U.S. specification)
Form
Liquid

Description

1-Octyne is widely used in organic reactions such as halogenation, hydration, oxidative cleavage, hydrogenation, hydrohalogenation, nitrile formation, polymerization and substitution reactions. It is used a precursor for the preparation of monohalogen substituted akenes or dihalogen substituted alkanes on treated with hydrogen halides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octyne is widely used in organic reactions such as halogenation, hydration, oxidative cleavage, hydrogenation, hydrohalogenation, nitrile formation, polymerization and substitution reactions. It is used a precursor for the preparation of monohalogen substituted akenes or dihalogen substituted alkanes on treated with hydrogen halides.

Solubility
Miscible with alcohol, ether and other organic solvents. Immiscible with water.

Notes
Incompatible with oxidizing agents. Keep away from heat, sparks, sources of ignition and flame.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text