Thermo Scientific Chemicals

Homovanillic acid, 98%, Thermo Scientific Chemicals

Catalog number: A12441.03
1 g, Each
Thermo Scientific Chemicals

Homovanillic acid, 98%, Thermo Scientific Chemicals

Catalog number: A12441.03
1 g, Each
Quantity
Catalog number: A12441.03
also known as A12441-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
306-08-1
IUPAC Name
2-(4-hydroxy-3-methoxyphenyl)acetic acid
Molecular Formula
C9H10O4
InChI Key
QRMZSPFSDQBLIX-UHFFFAOYSA-N
SMILES
COC1=CC(CC(O)=O)=CC=C1O
Assay (HPLC)
≥97.5%
Identification (FTIR)
Conforms (UK Sourced Material)
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to cream to pale brown to pale red
Comment
Material Sourced in UK and US

Description

Fluorimetric reagent and major catecholamine metaboliteHomovanillic acid is used as a reagent in the determination of oxidative enzymes. It is associated with dopamine levels in the brain. Its presence supports a diagnosis of neuroblastoma and malignant pheochromocytoma.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water,dimethyl sulfoxide, ethanol, methanol and ethyl acetate.

Notes
Light sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text