5-Amino-3-methylisoxazole, 98%, Thermo Scientific Chemicals
5-Amino-3-methylisoxazole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Amino-3-methylisoxazole, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number A12446.06
also known as A12446-06
Price (USD)/ Each
133.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
133.00
Add to cart
5-Amino-3-methylisoxazole, 98%, Thermo Scientific Chemicals
Catalog numberA12446.06
Price (USD)/ Each
133.00
-
Add to cart
Chemical Identifiers
CAS14678-02-5
IUPAC Name3-methyl-1,2-oxazol-5-amine
Molecular FormulaC4H6N2O
InChI KeyFNXYWHTZDAVRTB-UHFFFAOYSA-N
SMILESCC1=NOC(N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow to yellow to orange
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥98.0 to ≤102.0%
Assay (HPLC)≥98.0%
Identification (FTIR)Conforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only