Thermo Scientific Chemicals

1,8-Diazabicyclo[5.4.0]undec-7-ene, 98+%, Thermo Scientific Chemicals

Catalog number: A12449.14
25 g, Each
Thermo Scientific Chemicals

1,8-Diazabicyclo[5.4.0]undec-7-ene, 98+%, Thermo Scientific Chemicals

Catalog number: A12449.14
25 g, Each
Quantity
Catalog number: A12449.14
also known as A12449-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6674-22-2
IUPAC Name
2H,3H,4H,6H,7H,8H,9H,10H-pyrimido[1,2-a]azepine
Molecular Formula
C9H16N2
InChI Key
GQHTUMJGOHRCHB-UHFFFAOYSA-N
SMILES
C1CN=C2CCCCCN2C1
Form
Liquid
Refractive Index
1.5210-1.5240 @ 20?C
Identification (FTIR)
Conforms
Assay (GC)
≥98.0%
Appearance (Color)
Clear colorless to pale yellow

Description

1,8-Diazabicyclo[5.4.0]undec-7-ene is used in organic synthesis as a catalyst, a complexing ligand, and a non-nucleophilic base. It is used as a protecting agent for the synthesis of cephalosporin and as a catalyst for polyurethane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,8-Diazabicyclo[5.4.0]undec-7-ene is used in organic synthesis as a catalyst, a complexing ligand, and a non-nucleophilic base. It is used as a protecting agent for the synthesis of cephalosporin and as a catalyst for polyurethane.

Solubility
Miscible with water.

Notes
Incompatible with acids, strong oxidizing agents, acid anhydrides, acid chlorides and chloroformates.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text