4-Nitroimidazole, 97%, Thermo Scientific Chemicals
4-Nitroimidazole, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Nitroimidazole, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A12458.14
also known as A12458-14
Price (USD)/ Each
54.90
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
54.90
Add to cart
4-Nitroimidazole, 97%, Thermo Scientific Chemicals
Catalog numberA12458.14
Price (USD)/ Each
54.90
-
Add to cart
Chemical Identifiers
CAS3034-38-6
IUPAC Name5-nitro-1H-imidazole
Molecular FormulaC3H3N3O2
InChI KeyVYDWQPKRHOGLPA-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CN=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White to pale cream to pale yellow
Assay (HPLC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only