Thermo Scientific Chemicals

Diethyl 2,2-diethoxyethylphosphonate, 96%, Thermo Scientific Chemicals

Catalog number: A12461.09
10 g, Each
Thermo Scientific Chemicals

Diethyl 2,2-diethoxyethylphosphonate, 96%, Thermo Scientific Chemicals

Catalog number: A12461.09
10 g, Each
Quantity
Catalog number: A12461.09
also known as A12461-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7598-61-0
IUPAC Name
diethyl (2,2-diethoxyethyl)phosphonate
Molecular Formula
C10H23O5P
InChI Key
LUQYELQXRPNKRY-UHFFFAOYSA-N
SMILES
CCOC(CP(=O)(OCC)OCC)OCC
Appearance (Color)
Clear colorless
Refractive Index
1.4240-1.4300 @ 20?C
Assay (GC)
≥95.0%
Form
Liquid

Description

Diethyl 2,2-diethoxyethylphosphonate is used reactant for synthesis of α,β-alkenal derivatives by two-carbon homologation, lower rim-phosphonylated rexorcinol calix[4]arenes by condensation reactions, α-Phosphovinyl radicals via a radical trapping sequence, inhibitors of reverse transcriptase via 1,3-dipolar cycloadditions and for Friedel-Crafts reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl 2,2-diethoxyethylphosphonate is used reactant for synthesis of α,β-alkenal derivatives by two-carbon homologation, lower rim-phosphonylated rexorcinol calix[4]arenes by condensation reactions, α-Phosphovinyl radicals via a radical trapping sequence, inhibitors of reverse transcriptase via 1,3-dipolar cycloadditions and for Friedel-Crafts reactions.

Solubility
Soluble in chloroform and dichloromethane. Not miscible or difficult to mix in water.

Notes
Keep container tightly sealed. It is hygroscopic in nature. It is sensitive to moisture. Store under dry inert gas. Protect from humidity and water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text