Thermo Scientific Chemicals

2,4-Dichlorophenoxyacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: A12467.30
250 g, Each
Thermo Scientific Chemicals

2,4-Dichlorophenoxyacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: A12467.30
250 g, Each
Quantity
Catalog number: A12467.30
also known as A12467-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
94-75-7
IUPAC Name
2-(2,4-dichlorophenoxy)acetic acid
Molecular Formula
C8H6Cl2O3
InChI Key
OVSKIKFHRZPJSS-UHFFFAOYSA-N
SMILES
OC(=O)COC1=CC=C(Cl)C=C1Cl
Appearance (Color)
White to pale cream to pale brown
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
134.0-142.0?C

Description

2,4-Dichlorophenoxyacetic acid is used as a supplement in plant cell culture media. It is a synthetic auxin, which finds application as a plant growth regulator. Further, it is used to increase latex output of old rubber trees.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dichlorophenoxyacetic acid is used as a supplement in plant cell culture media. It is a synthetic auxin, which finds application as a plant growth regulator. Further, it is used to increase latex output of old rubber trees.

Solubility
Slightly soluble in water.

Notes
Moisture and light-sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text