Formanilide, 98%, Thermo Scientific Chemicals
Formanilide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Formanilide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number A12472.22
also known as A12472-22
Price (USD)/ Each
54.70
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
54.70
Add to cart
Formanilide, 98%, Thermo Scientific Chemicals
Catalog numberA12472.22
Price (USD)/ Each
54.70
-
Add to cart
Chemical Identifiers
CAS103-70-8
IUPAC NameN-phenylformamide
Molecular FormulaC7H7NO
InChI KeyDYDNPESBYVVLBO-UHFFFAOYSA-N
SMILESO=CNC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to grey
Melting Point (clear melt)45-50?C
FormCrystals or powder or crystalline powder or fused/lumpy solid
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only