Thermo Scientific Chemicals

Formanilide, 98%, Thermo Scientific Chemicals

Catalog number: A12472.36
500 g, Each
Thermo Scientific Chemicals

Formanilide, 98%, Thermo Scientific Chemicals

Catalog number: A12472.36
500 g, Each
Quantity
Catalog number: A12472.36
also known as A12472-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
103-70-8
IUPAC Name
N-phenylformamide
Molecular Formula
C7H7NO
InChI Key
DYDNPESBYVVLBO-UHFFFAOYSA-N
SMILES
O=CNC1=CC=CC=C1
Appearance (Color)
White to cream to grey
Melting Point (clear melt)
45-50?C
Form
Crystals or powder or crystalline powder or fused/lumpy solid
Assay (GC)
≥97.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text