Ammonium iron(II) sulfate hexahydrate, 99%, Thermo Scientific Chemicals
Ammonium iron(II) sulfate hexahydrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium iron(II) sulfate hexahydrate, 99%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
500 g
Catalog number A12473.36
also known as A12473-36
Price (USD)/ Each
61.65
Online exclusive
67.90 
Save 6.25 (9%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
61.65
Online exclusive
67.90 
Save 6.25 (9%)
Add to cart
Ammonium iron(II) sulfate hexahydrate, 99%, Thermo Scientific Chemicals
Catalog numberA12473.36
Price (USD)/ Each
61.65
Online exclusive
67.90 
Save 6.25 (9%)
-
Add to cart
Chemical Identifiers
CAS7783-85-9
IUPAC Nameλ²-iron(2+) diammonium hexahydrate disulfate
Molecular FormulaFeH20N2O14S2
InChI KeyMQLVWQSVRZVNIP-UHFFFAOYSA-L
SMILES[NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid
Assay (Redox Titration)≥98.5 to ≤101.5%
Appearance (Color)White to green or pale blue
Ammonium iron(II) sulfate hexahydrate is widely utilized in analytical chemistry for titration purposes since it is affected lesser by oxygen in the air than iron(II) sulpahte. It is employed to measure high doses of gamma rays by using Fricke's dosemeter. It is involved in a variety of other applications from nanomaterials to general redox reactions. It is used in the medical field as an antianemic agent for treatment of iron deficiency.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water. Insoluble in alcohol.

Notes
Air and light sensitive. Incompatible with strong acids and oxidizing agents.
RUO – Research Use Only

General References:

  1. Uchida, M.; Qazi, S.; Edwards, E.; Douglas, T. Use of Protein Cages as a Template for Confined Synthesis of Inorganic and Organic Nanoparticles. Methods Mol. Biol. 2014, 1252, 17-25.
  2. Karvinen, A.; Lehtinen, L.; Kankaala, P. Variable Effects of Iron (Fe (III)) Additions on Potential Methane Production in Boreal Lake Littoral Sediments. Wetlands 2015, 35 (1), 137-146.