Thermo Scientific Chemicals

5-Hydroxymethyl-2-furaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: A12475.06
5 g, Each
Thermo Scientific Chemicals

5-Hydroxymethyl-2-furaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: A12475.06
5 g, Each
Quantity
Catalog number: A12475.06
also known as A12475-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
67-47-0
IUPAC Name
5-(hydroxymethyl)furan-2-carbaldehyde
Molecular Formula
C6H6O3
InChI Key
NOEGNKMFWQHSLB-UHFFFAOYSA-N
SMILES
OCC1=CC=C(O1)C=O
Form
Crystals or powder or crystalline powder or lumps or fused solid or clear liquid as melt
Identification (FTIR)
Conforms
Melting Point (clear melt)
27.0-37.0?C
Appearance (Color)
Pale cream to cream to yellow to pale brown
Water Content (Karl Fischer Titration)
≤1.5%

Description

5-Hydroxymethyl-2-furaldehyde is used as an intermediate of OMBF (5,5'-oxydimethylenebis(2- furfural)), crown ethers. It is used in organic synthesis to prepare dialdehydes, glycols, ethers, amino alcohols and acetals. It acts as an antioxidant found in some fruit juices and caramel products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Hydroxymethyl-2-furaldehyde is used as an intermediate of OMBF (5,5′-oxydimethylenebis(2- furfural)), crown ethers. It is used in organic synthesis to prepare dialdehydes, glycols, ethers, amino alcohols and acetals. It acts as an antioxidant found in some fruit juices and caramel products.

Solubility
Soluble in water, alcohol, ethyl acetate, acetone, dimethylformamide, benzene, ether and chloroform.

Notes
Air and light sensitive. Keep the container tightly closed. Incompatible with strong bases, strong oxidizing agents and strong reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text