Informational:Taking your work to the clinic? Check out CTS Hibernate -A Medium »
Gibco™

Hibernate™-A Medium

Catalog number: A1247501
Have Questions?
Gibco™

Hibernate™-A Medium

Catalog number: A1247501
Have Questions?
Catalog Number
A1247501
Unit Size
500 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A1247501500 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Hibernate Medium is a CO2-independent nutrient medium for the maintenance of neural cells, tissue, and tissue slices. Hibernate-A Medium is optimized for adult and postnatal cells or tissue in ambient carbon dioxide levels.

Hibernate Medium enables neurons to be maintained in ambient CO2 levels for up to 48 hours. This provides scientists with greater convenience and control while running bench-top live cell experiments, such as microscopy, flow cytometry, and other physiology studies. Hibernate Medium also allows for the preservation of viable brain tissue before processing for up to a month with refrigeration and maintenance of tissue slices.

Hibernate Medium may be combined with B-27 Supplements to yield a complete medium.

For other applications, we provide alternative formulations of Hibernate Medium and B-27 Supplements.

For established protocols using Hibernate Medium, access our Gibco Neurobiology Protocol Handbook.

View publications using Gibco neurobiology products at our Gibco CellCite.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type
Neural Cells
Classification
Serum-free
Culture Type
Mammalian Cell Culture
Form
Liquid
Product Type
Cell Culture Medium
Serum Level
Serum-free
Shelf Life
24 Months
Sterility
Sterile
With Additives
Phenol Red
Product Line
Hibernate™
Shipping Condition
Ambient
Tested For
Cytotoxicity, Endotoxin, Sterility, pH, Osmolality

Contents & Storage

Store in refrigerator (2–8°C) and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text