Thermo Scientific Chemicals

Diethyl carbonate, 99%, Thermo Scientific Chemicals

Catalog number: A12477.0F
2500 mL, Each
Thermo Scientific Chemicals

Diethyl carbonate, 99%, Thermo Scientific Chemicals

Catalog number: A12477.0F
2500 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A12477.0F
also known as A12477-0F
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
105-58-8
IUPAC Name
diethyl carbonate
Molecular Formula
C5H10O3
InChI Key
OIFBSDVPJOWBCH-UHFFFAOYSA-N
SMILES
CCOC(=O)OCC
Assay (GC)
≥99.0%
Refractive Index
1.3835-1.3855 @ 20?C
Appearance (Color)
Clear colorless
Identification (FTIR)
Conforms
Form
Liquid

Description

Diethyl carbonate is esters, beta-enamino esters, carbamates and unsymmetrical alkyl carbonates. It is an active component of electrolytes used in lithium which is used as a solvent for cellulose ethers, nitro cellulose, natural and synthetic resin and in erythromycin intramuscular injections. It is also used in the synthesis of 3-ethyl-4-methyl-5-phenyl-3H-oxazol-2-one, phenobarbital, pyrethrum batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl carbonate is esters, beta-enamino esters, carbamates and unsymmetrical alkyl carbonates. It is an active component of electrolytes used in lithium which is used as a solvent for cellulose ethers, nitro cellulose, natural and synthetic resin and in erythromycin intramuscular injections. It is also used in the synthesis of 3-ethyl-4-methyl-5-phenyl-3H-oxazol-2-one, phenobarbital, pyrethrum batteries.

Solubility
Miscible with diehtyl ether, ethanol and chloroform.Immiscible with water.

Notes
Store in cool place. Moisture sensitive. Incompatible with strong acids, oxidizing agents, strong bases and reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text