Thermo Scientific Chemicals

Cyanoacetamide, 99%, Thermo Scientific Chemicals

Catalog number: A12482.22
100 g, Each
Thermo Scientific Chemicals

Cyanoacetamide, 99%, Thermo Scientific Chemicals

Catalog number: A12482.22
100 g, Each
Quantity
Catalog number: A12482.22
also known as A12482-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
107-91-5
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
118-123?C
Assay (GC)
≥98.5%
Appearance (Color)
White to pale cream
Identification (FTIR)
Conforms

Description

Cyanoacetamide was used in spectrofluorimetric method for the determination of some antihistaminic H1 receptor antagonist drugs such as ebastine, cetirizine dihydrochloride and fexofenadine hydrochloride. It may be used in fluorometric determination of 3,4-dihydroxyphenylalanine. It was used as post column fluorigenic derivatizing agent for the bio-analysis and pharmacokinetics of chitosan ester in rabbit serum. Cyanoacetamide is the starting reagent for vitamin B6 synthesis. It reacts with reducing carbohydrates in borate buffer to give intense fluorescence andis useful for the automated analysis of carbohydrates as borate complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyanoacetamide was used in spectrofluorimetric method for the determination of some antihistaminic H1 receptor antagonist drugs such as ebastine, cetirizine dihydrochloride and fexofenadine hydrochloride. It may be used in fluorometric determination of 3,4-dihydroxyphenylalanine. It was used as post column fluorigenic derivatizing agent for the bio-analysis and pharmacokinetics of chitosan ester in rabbit serum. Cyanoacetamide is the starting reagent for vitamin B6 synthesis. It reacts with reducing carbohydrates in borate buffer to give intense fluorescence andis useful for the automated analysis of carbohydrates as borate complexes.

Solubility
Soluble in water.

Notes
Store under cool dry place. Ensure good ventilation. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text