Bismuth carbonate oxide, Thermo Scientific Chemicals
Bismuth carbonate oxide, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bismuth carbonate oxide, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number A12483.18
also known as A12483-18
Price (USD)/ Each
33.90
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
33.90
Add to cart
Bismuth carbonate oxide, Thermo Scientific Chemicals
Catalog numberA12483.18
Price (USD)/ Each
33.90
-
Add to cart
Chemical Identifiers
CAS5892-10-4
IUPAC Name({[(oxobismuthanyl)oxy]carbonyl}oxy)bismuthanone
Molecular FormulaCBi2O5
InChI KeyMGLUJXPJRXTKJM-UHFFFAOYSA-L
SMILESO=[Bi]OC(=O)O[Bi]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on Drying≤ 1.0%
Formpowder
Appearance (Color)White to pale cream or pale yellow
Complexometric TitrationBismuth Bi content (dry weight): 80.0 - 82.5% (theory 81.96%)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only