Methyl alpha-D-glucopyranoside, 98%, Thermo Scientific Chemicals
Methyl alpha-D-glucopyranoside, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl alpha-D-glucopyranoside, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number A12484.36
also known as A12484-36
Price (USD)/ Each
144.00
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
144.00
Add to cart
Methyl alpha-D-glucopyranoside, 98%, Thermo Scientific Chemicals
Catalog numberA12484.36
Price (USD)/ Each
144.00
-
Add to cart
Chemical Identifiers
CAS97-30-3
IUPAC Name2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
Molecular FormulaC7H14O6
InChI KeyHOVAGTYPODGVJG-UHFFFAOYNA-N
SMILESCOC1OC(CO)C(O)C(O)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Melting Point165.0-170.0?C
FormCrystals or powder or crystalline powder
Water Content (Karl Fischer Titration)≤0.5%
Optical Rotation+159? ± 3? (c=10 in water)
View more
Methyl alpha-D-glucopyranoside is a biochemical used to vary echo decay times in a study that assessed the effects of cryoprotection on the structure and activity of p21ras. It is also used in a study to investigate saccharide-mediated protection of chaotropic-induced deactivation of concanavalin A.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl alpha-D-glucopyranoside is a biochemical used to vary echo decay times in a study that assessed the effects of cryoprotection on the structure and activity of p21ras. It is also used in a study to investigate saccharide-mediated protection of chaotropic-induced deactivation of concanavalin A.

Solubility
Soluble in water. Insoluble in ether

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature. Store under dry inert gas.
RUO – Research Use Only

General References:

  1. D N Figlas.; H R Arias.; A Fernández.; D M Alperin. Dramatic saccharide-mediated protection of chaotropic-induced deactivation of concanavalin A. Archives of Biochemistry and Biophysics. 1997, 340, (1), 154-158.
  2. C J Halkides.; C T Farrar.; D J Singel. The effects of cryoprotection on the structure and activity of p21 ras: implications for electron spin-echo envelope modulation spectroscopy. Journal of Magnetic Resonance. 1998, 134 (1), 142-153.