Tris(4-chlorophenyl)phosphine, 97%, Thermo Scientific Chemicals
Tris(4-chlorophenyl)phosphine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tris(4-chlorophenyl)phosphine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number A12487.14
also known as A12487-14
Price (USD)/ Each
918.65
Online exclusive
1020.00 
Save 101.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
918.65
Online exclusive
1020.00 
Save 101.35 (10%)
Add to cart
Tris(4-chlorophenyl)phosphine, 97%, Thermo Scientific Chemicals
Catalog numberA12487.14
Price (USD)/ Each
918.65
Online exclusive
1020.00 
Save 101.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1159-54-2
IUPAC Nametris(4-chlorophenyl)phosphane
Molecular FormulaC18H12Cl3P
InChI KeyIQKSLJOIKWOGIZ-UHFFFAOYSA-N
SMILESClC1=CC=C(C=C1)P(C1=CC=C(Cl)C=C1)C1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder or powder
Assay (GC)≥96.0%
Melting Point96-103°C
Appearance (Color)White to cream or pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only