Thermo Scientific Chemicals

Methyl dodecanoate, 99%, Thermo Scientific Chemicals

Catalog number: A12492.06
5 g, Each
Thermo Scientific Chemicals

Methyl dodecanoate, 99%, Thermo Scientific Chemicals

Catalog number: A12492.06
5 g, Each
Quantity
Catalog number: A12492.06
also known as A12492-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
111-82-0
IUPAC Name
methyl dodecanoate
Molecular Formula
C13H26O2
InChI Key
UQDUPQYQJKYHQI-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCC(=O)OC
Form
Liquid
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Refractive Index
1.4290-1.4340 @ 20?C
Appearance (Color)
Clear colorless to pale yellow

Description

Methyl dodecanoate used in production of biodiesel. It has been used in selective synthesis of the secondary amide surfactant, N-methyl lauroylethanolamide. It is also utilized as emollients, perfuming agents skin conditioning.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl dodecanoate used in production of biodiesel. It has been used in selective synthesis of the secondary amide surfactant, N-methyl lauroylethanolamide. It is also utilized as emollients, perfuming agents skin conditioning.

Solubility
Not miscible in water. Soluble in alcohol, dipropylene glycol.

Notes
Store at room temperature. Ensure good ventilation.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text