Butyramide, 98%, Thermo Scientific Chemicals
Butyramide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Butyramide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
100 g
500 g
Catalog number A12494.18
also known as A12494-18
Price (USD)/ Each
45.70
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
45.70
Add to cart
Butyramide, 98%, Thermo Scientific Chemicals
Catalog numberA12494.18
Price (USD)/ Each
45.70
-
Add to cart
Chemical Identifiers
CAS541-35-5
IUPAC Namebutanamide
Molecular FormulaC4H9NO
InChI KeyDNSISZSEWVHGLH-UHFFFAOYSA-N
SMILESCCCC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or flakes
Assay (GC)≥97.5%
Melting Point (clear melt)109.0-115.0?C
Appearance (Color)White to cream
Identification (FTIR)Conforms
Butyramide was used in the synthesis of hydroxamic acids, electrorheological fluids and β-amodoorganotin compounds. It was used as substrate of (+)-γ-lactamase to develop a microreactor to study enzyme stability, activity, kinetics and substrate specificity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Butyramide was used in the synthesis of hydroxamic acids, electrorheological fluids and β-amodoorganotin compounds. It was used as substrate of (+)-γ-lactamase to develop a microreactor to study enzyme stability, activity, kinetics and substrate specificity.

Solubility
Soluble in water (170 g/L).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. J.Raj; A.Seth; S.Prasad; T.C.Bhalla. Bioconversion of butyronitrile to butyramide using whole cells of Rhodococcus rhodochrous PA-34. Applied Microbiology and Biotechnology. 2007, 74, (3)535-539
  2. Anne Marie Hickey; Bongkot Ngamsom; Charlotte Wiles; Gillian M.Greenway; Paul Watts; Jennifer A.Littlechild. A microreactor for the study of biotransformations by a cross-linked gamma-lactamase enzyme. Biotechnology Journal. 2009, 4, (4)510-516