3'-Aminoacetophenone, 97%, Thermo Scientific Chemicals
3'-Aminoacetophenone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3'-Aminoacetophenone, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1000 g
Catalog number A12503.30
also known as A12503-30
Price (USD)/ Each
112.00
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
112.00
Add to cart
3'-Aminoacetophenone, 97%, Thermo Scientific Chemicals
Catalog numberA12503.30
Price (USD)/ Each
112.00
-
Add to cart
Chemical Identifiers
CAS99-03-6
IUPAC Name1-(3-aminophenyl)ethan-1-one
Molecular FormulaC8H9NO
InChI KeyCKQHAYFOPRIUOM-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=CC(N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Melting Point95-100?C
Appearance (Color)Cream or yellow to brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only