Thermo Scientific Chemicals

L-Glutamic acid hydrochloride, 99%

Catalog number: A12505.30
250 g, Each
Thermo Scientific Chemicals

L-Glutamic acid hydrochloride, 99%

Catalog number: A12505.30
250 g, Each
Quantity
Catalog number: A12505.30
also known as A12505-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
138-15-8
IUPAC Name
hydrogen 2-aminopentanedioic acid chloride
Molecular Formula
C5H10ClNO4
InChI Key
RPAJSBKBKSSMLJ-UHFFFAOYNA-N
SMILES
[H+].[Cl-].NC(CCC(O)=O)C(O)=O
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (Titration ex Chloride)
≥98.5 to ≤101.5% (non-U.S. specification)
Assay from Suppliers CofA
≥98.5% (U.S. specification)
Comment
Ammonium (NH4): ≤200 ppm (non-U.S. specification)

Description

L-Glutamic acid hydrochloride is used as a digestive aid. It is used in cell culture as a component of MEM non-essential amino acids solution. It has been used as a nitrogen source in the culture of Aspergillus fumigatus NRRL 2436 for fumagillin production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Glutamic acid hydrochloride is used as a digestive aid. It is used in cell culture as a component of MEM non-essential amino acids solution. It has been used as a nitrogen source in the culture of Aspergillus fumigatus NRRL 2436 for fumagillin production.

Solubility
Soluble in water (490 g/L).

Notes
Hygroscopic. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text