Thermo Scientific Chemicals

1-Nonanol, 99%, Thermo Scientific Chemicals

Catalog number: A12510.36
500 g, Each
Thermo Scientific Chemicals

1-Nonanol, 99%, Thermo Scientific Chemicals

Catalog number: A12510.36
500 g, Each
Quantity
Catalog number: A12510.36
also known as A12510-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
143-08-8
IUPAC Name
nonan-1-ol
Molecular Formula
C9H20O
InChI Key
ZWRUINPWMLAQRD-UHFFFAOYSA-N
SMILES
CCCCCCCCCO
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥98.5%
Refractive Index
1.4320-1.4350 @ 20?C

Description

The primary use of nonanol is in the manufacture of artificial lemon oil. 1-Nonanol is mainly used in man-made rose essence, plasticizer, surfactant. 1-Nonanol was used as a reference material in the determination of airborne spore levels. 1-Nonanol inhibited the bacterial luciferase (BL) reaction in a dose-dependent manner.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The primary use of nonanol is in the manufacture of artificial lemon oil. 1-Nonanol is mainly used in man-made rose essence, plasticizer, surfactant. 1-Nonanol was used as a reference material in the determination of airborne spore levels. 1-Nonanol inhibited the bacterial luciferase (BL) reaction in a dose-dependent manner.

Solubility
Soluble in water (1 g/L) at 20°C, ether, ethanol, acetone, chloroform, and mineral oil.

Notes
Store at room temperature. Ensure good ventilation.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text