Thermo Scientific Chemicals

Glycolic acid, 98%, Thermo Scientific Chemicals

Catalog number: A12511.36
500 g, Each
Thermo Scientific Chemicals

Glycolic acid, 98%, Thermo Scientific Chemicals

Catalog number: A12511.36
500 g, Each
Quantity
Catalog number: A12511.36
also known as A12511-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
79-14-1
IUPAC Name
2-hydroxyacetic acid
Molecular Formula
C2H4O3
InChI Key
AEMRFAOFKBGASW-UHFFFAOYSA-N
SMILES
OCC(O)=O
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
≤2%

Description

Glycolic acid is used for acne and anti-aging treatment. It is also employed in the textile industry as a dyeing and tanning agent. It is used in emulsion polymers, solvents and additives for ink and paints. It is also useful as an intermediate for organic synthesis such as redox reactions, esterification and long chain polymerization. It acts as a monomer in the preparation of polyglycolic acid and other biocompatible copolymers. Further, it is used as a preservative and flavoring agent in organic synthesis. It is also used in adhesives and plastics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycolic acid is used for acne and anti-aging treatment. It is also employed in the textile industry as a dyeing and tanning agent. It is used in emulsion polymers, solvents and additives for ink and paints. It is also useful as an intermediate for organic synthesis such as redox reactions, esterification and long chain polymerization. It acts as a monomer in the preparation of polyglycolic acid and other biocompatible copolymers. Further, it is used as a preservative and flavoring agent in organic synthesis. It is also used in adhesives and plastics.

Solubility
Soluble in water, methanol, diethyl ether, acetone, acetic acid and ethyl acetate.

Notes
Hygroscopic. Incompatible with bases, oxidizing agents and reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text