Thermo Scientific Chemicals

Thiophene-2-carboxylic acid, 99%, Thermo Scientific Chemicals

Catalog number: A12514.22
100 g, Each
Thermo Scientific Chemicals

Thiophene-2-carboxylic acid, 99%, Thermo Scientific Chemicals

Catalog number: A12514.22
100 g, Each
Quantity
Catalog number: A12514.22
also known as A12514-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
527-72-0
IUPAC Name
thiophene-2-carboxylic acid
Molecular Formula
C5H4O2S
InChI Key
QERYCTSHXKAMIS-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=CS1
Appearance (Color)
White to cream to pale brown
Form
Crystalline powder
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Melting Point (clear melt)
126-132?C
Assay (GC)
≥98.5%

Description

A hypoglycemic, anti-lipolytic agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text